Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

American Scientific Plastic Handheld Magnifier
SDPSCI_ED

Catalog No. S02457
Change view
Click to view available options
:
ProductType: Hand Magnifier
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S02457 ProductType: Hand Magnifier
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S02457 Supplier American Scientific Supplier No. 645750
This item is not returnable. View return policy

This dual lens Plastic Handheld Magnifier is economical and sturdy.

  • Magnification: 3X and 6X
  • Approx. 50 mm (2 in.) Dia.
  • Great for classroom use

Specifications

Product Type Magnifier, Handheld
Description Dual Lens
Material Plastic
Diameter (Englis) Lens 2 in.
Diameter (Metric) Lens 50 mm
Magnification 3, 6x
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.