Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Plastic Petri Dish Set
GREENER_CHOICESCI_ED

Catalog No. S81668 Shop All Supertek Scientific Products
Click to view available options
Diameter (Metric):
90 mm

Catalog No. S81668

Supplier: Supertek™ CH10928S3

Only null left
Add to Cart
Add to Cart

Discover the fun of microbiology with plastic Petri Dishes and Nutreint Agar.

  • Discover the fun of microbiology
  • Three 90 mm Petri dishes and one bag of Nutrient Agar included
  • Nutrient Agar is used as a culture medium to cultivate various household bacteria
  • Detailed agar preparation instructions included
  • Dishes are polystyrene and sterilized
  • Ages 7+

Specifications

Bottom Shape Flat
Color Clear
Material Polystyrene
Sterility Sterile
Surface Treatment Non-treated
Type Petri Dish
Includes Set of 3 Dishes with Agar
Height (Metric) 1.5cm
Diameter (Metric) 90 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.