Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Plastic Tight Head Drums
SDP

Catalog No. p-4907981
Click to view available options
Color:
Blue
Natural
Certifications/Compliance:
1H1/Y1.9/150
UN 1H1/Y1.8/100
UN 1H1/Y1.9/100
UN 1H1/Y1.9/150
Diameter (English) Outer:
14.2 in.
19.25 in.
19.5 in.
23.3 in.

High-density polyethylene containers for transport of hazardous solids or liquids

  • Made of chemical-resistant high-density polyethylene
  • Tight-head drums for transport of hazardous liquids

Specifications

Certifications/Compliance UN 1H1/Y1.8/100
Product Type Plastic Tight Head Drums
Capacity (English) 15 gal.
Capacity (Metric) 56.78 L
Color Blue
For Use With (Application) For Transport of Hazardous Solids or Liquids
Material Plastic
Freight Class 50
Head Style Tight
Height (English) 26.25 in.
Height (Metric) 66.67 cm
Diameter (English) Outer 14.2 in.
Diameter (Metric) Outer 36.06 mm
Weight (English) 6.5 lb.
Weight (Metric) 2.94 Kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.