Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plastin L Antibody (29G10), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32617450UL
Description
Plastin L Monoclonal antibody specifically detects Plastin L in Human samples. It is validated for Flow Cytometry, ELISASpecifications
Plastin L | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA | |
29G10 | |
Flow Cytometry 1:50-1:200, ELISA | |
CP64, DKFZp781A23186, FLJ25423, FLJ26114, FLJ39956, LC64PPLS2bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)), LCP-1, LPL, L-plastin, L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P), Lymphocyte cytosolic protein 1, lymphocyte cytosolic protein 1 (L-plastin), Lymphocyte cytosolic protein-1 (plasmin), plastin 2, plastin-2 | |
A synthesized peptide derived from Human Plastin L [UniProt P13796] | |
50 μL | |
Cancer, Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction | |
3936 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction