Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Plate Seal for Pierce™ 96-well Microdialysis Plates PROMO
Click to view available options
:
Thermo Scientific™ Plate Seal for Pierce™ 96-well Microdialysis Plates
Description
Thermo Scientific Pierce Plate Seal for Pierce 96-well Microdialysis Plates is an adhesive coated plastic sheet, perforated for use with an 8-well strip microdialysis plate. Sufficient For: 12 x 8 microdialysis plate wells.
These plate seals are made of high-quality polypropylene and help protect samples from contamination and evaporation, especially during long-term storage.
Specifications
Specifications
Sufficient For | 12 x 8 microdialysis plate wells |
Content And Storage | Upon receipt store at room temperature. Product shipped at ambient temperature. |
Format | Microdialysis Plates |
Product Line | Slide-A-Lyzer™ |
Quantity | 1 Sheet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction