Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platelet Monoclonal Antibody (AIP21)
GREENER_CHOICE

Catalog No. MA170084
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA170084 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA170084 Supplier Invitrogen™ Supplier No. MA170084
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MA1-70084 detects Platelet in mouse samples. MA1-70084 has been successfully used in flow cytometry procedures. The MA1-70084 immunogen is platelets.

Platelets are nonproliferative and terminally differentiated cells. Platelets offer an attractive model system to study the various biochemical events leading to structural and functional alterations in activated cells. When platelets are exposed to stimuli, they are activated, undergo a dramatic shape change, adhere to each other, and aggregate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Platelet
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AIP21
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias Thrombocytes
Host Species Rat
Immunogen Platelet.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.