Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platelet Monoclonal Antibody (AIP21), FITC

Catalog No. PIMA517901
Change view
Click to view available options
Quantity:
100 μg
300 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517901 300 μg
PIMA517900 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517901 Supplier Invitrogen™ Supplier No. MA517901
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Platelet Monoclonal Antibody for Flow

Platelets are nonproliferative and terminally differentiated cells. Platelets offer an attractive model system to study the various biochemical events leading to structural and functional alterations in activated cells. When platelets are exposed to stimuli, they are activated, undergo a dramatic shape change, adhere to each other, and aggregate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Platelet
Applications Flow Cytometry
Classification Monoclonal
Clone AIP21
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Alias Thrombocytes
Host Species Rat
Immunogen Mouse Platelets.
Purification Method Affinity chromatography
Quantity 300 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.