Learn More
Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane
Description
Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst.Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane is used as a catalyst in hydrosilylation reaction for the preparation of trans-beta-silanes and in reductive amination reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst
Specifications
Specifications
| Color | Orange |
| Density | 0.984 g/mL |
| Flash Point | >110°C (230°F) |
| Quantity | 5 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Miscible with toluene,xylene,isopropyl alcohol,double-vinyl silicone oil and vinyl terminated poly(dimethylsiloxane). Immiscible with water. |
| IUPAC Name | ethenyl[(ethenyldimethylsilyl)oxy]dimethylsilane platinum |
| Formula Weight | 474.68 |
| Physical Form | Liquid |
| Chemical Name or Material | Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, Soln. in vinyl terminated polydimethylsiloxane |
Safety and Handling
| Hazard Category | H335 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.