Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Platinum On Carbon
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 g
Description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Platinum on carbon (Pt/C) can be used as a catalyst for a variety of applications, such as:hydrosilylation reaction for the production of molecules with silicon-carbon bondsdirect alcohol fuel cells (DAFCs)an electrocatalyst in hydrogen evolution reaction (HER)formation of nanocatalyst for polymer electrolyte fuel cells
Specifications
Specifications
| Molecular Formula | C16H14CoN2O2 -+ xH2O |
| Linear Formula | [CH2N=CHC6H4-2-(O)]2Co-+xH2O |
| MDL Number | MFCD00149054 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 325.23 g/mol |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction