Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Platinum Catalytic Static Mixer 316L 6Mm Dia X 150Mm

Catalog No. 9196751EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-967-51EA 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-967-51EA Supplier Sigma Aldrich Supplier No. 9196751EA
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Platinum Catalytic Static Mixer 316L 6mm dia x 150mm is a general hydrogenation catalyst that exhibits moderate to high activity with good selectivity toward carbonyl over alkene reduction. This CSM has a long lifetime.Core: 3D printed 316L stainless steelCatalyst: electroplated PtShape: cylindricalDimensions: 6mm diameter x 150m mLengthCatalytic static mixers are a novel immobilized catalyst system, based on 3D printed mixer scaffolds containing a catalytic active layer. A variety of different active catalysts can be deposited on the metal mixer, allowing employment in many different classes of chemical reactions, such as hydrogenations, oxidations, C-C couplings and many more.For additional information, including a general operational guide, click hereContinuous flow hydrogenations using novel catalytic static mixers inside a tubular reactorReductive aminations using a 3D printed supported metal(0) catalyst systemThe use of catalytic static mixers for continuous flow gas-liquid and transfer hydrogenations in organic synthesisCatalytic Static Mixers for the Continuous Flow Hydrogenation of a Key Intermediate of Linezolid (Zyvox)

Specifications

Molecular Formula C6H10F3NO2
Quantity 500 mg
Molecular Weight (g/mol) 185.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.