Alfa Aesar™ Platinum foil, 0.1mm thick, 99.99% (metals basis), Alfa Aesar™, Premion™

Manufacturer: Alfa Aesar™ 11509FF

Quantity 25 x 25mm

 View more versions of this product

Catalog No. AA11509FF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-06-4
    MDL Number MFCD00011179
    Chemical Name or Material Platinum

    Specifications

    Quantity 25 x 25mm
    Density 21.45g/cm3 at 20°C
    EINECS Number 231-116-1
    Physical Form Foil, 0.1mm (0.004 in.) thick
    Assay Percent Range 99.99% (Metals basis)
    MDL Number MFCD00011179
    CAS 7440-06-4
    Odor Odorless
    Appearance Silvery white foil
    Chemical Name or Material Platinum

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit