Learn More
Invitrogen™ Platinum™ Hot Start PCR Master Mixes (2X)
Description
Invitrogen Platinum Hot Start PCR Master Mix, available in colorless and green formats, offers Platinum Taq DNA polymerase in an optimized PCR buffer with magnesium and dNTPs for convenient PCR setup. It retains all the features of Platinum Taq DNA Polymerase: specificity, robustness, and reliability.
Platinum Green Hot Start PCR Master Mix additionally contains a density reagent and two tracking dyes for direct loading of PCR products on gels.
Platinum Hot Start PCR Master Mix provides a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs, thus reducing the number of pipetting steps during PCR reaction setup. It is supplied with a separate vial of Platinum GC Enhancer, formulated for specific amplification and improved yields of GC-rich targets.
Platinum Taq DNA Polymerase is a recombinant Taq DNA polymerase complexed with proprietary antibodies that inhibit enzyme activity at room temperature, reducing nonspecific target amplification at lower temperatures. The antibodies dissociate during the initial PCR denaturation step, and DNA polymerase regains its full activity. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template-dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5' to 3' exonuclease activity.
Features
- High specificity and increased yields with antibody-mediated hot-start PCR
- Convenient room temperature reaction setup
- Versatile formulation for broad range of amplicons
- Direct gel loading with green PCR master mix
Applications
- Hot Start PCR
- Genotyping
- Gene expression profiling
- Next-generation sequencing
Notes
- Platinum II Taq Hot-start DNA Polymerase is recommended superior PCR performance.
- For applications that require PCR product analysis by absorbance or fluorescence excitation, colorless format is recommended.
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | • Platinum Hot Start PCR 2X Master Mix, 4 x 1.25 mL • Platinum GC Enhancer, 2 x 1.25 mL • Nuclease-free water, 4 x 1.25 mL Store at -20°C in a non-frost-free freezer. |
| Format | Tube |
| GC-Rich PCR Performance | Medium |
| Polymerase | Platinum Taq DNA Polymerase |
| Reaction Speed | Standard |
| Label or Dye | Unlabeled |
| Product Type | Hot Start PCR Master Mix |
| Quantity | 200 reactions |
| Shipping Condition | Dry Ice |
| Show More |
Frequently Asked Questions (FAQs)
We have the following recommendations:
Increase the extension time from 1 min/kb to 1.5 min/kb.
Vary total PCR cycles from 20-40; 35 cycles is typical.
Try denaturing at 95 degrees C for 45 seconds.
GC-rich or problematic targets work better with MgSO4 instead of MgCl2.
For GC-rich templates, test higher annealing temperatures starting with the temperature that is equal to your primer Tm and in increments of 2 degrees C for up to six different temperatures.
Note: Addition of Platinum GC Enhancer solution enhances the amplification of GC-rich and problematic sequences.
The final concentration is 1.5 mM MgCl2.
We recommend using the GC enhancer for targets with >65% GC sequences. We recommend that you have a final concentration of 20% of the GC enhancer in the reaction mix. There is enough GC enhancer included in the kit to add it to every PCR reaction.
Yes, we offer several Platinum Taq DNA polymerases and their master mixes, supplemented with tracking dyes for direct loading of PCR products on gels:
- Platinum II Hot-Start Green PCR Master Mix (2X)
- Platinum SuperFi Green DNA Polymerase and Platinum SuperFi Green PCR Master Mix
- Platinum Taq Green Hot-Start DNA Polymerase and Platinum Green Hot-Start PCR Master Mix (2X)
- DreamTaq Green Hot-Start DNA Polymerase and DreamTaq Hot-Start Green PCR Master Mix
- Phusion Green Hot-Start II High-Fidelity DNA Polymerase and Phusion Green Hot-Start II High-Fidelity PCR Master Mix.
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.