Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(II) 2,4-pentanedionate, Pt 48.0% min

Catalog No. p-4407705
Click to view available options
Quantity:
1 g
5 g

CAS: 15170-57-7 | C10H14O4Pt | 393.30 g/mol

Used for photoactivated hydrosilylation reactions between vinyl and silane groups; in the deposition of platinum metal and in the preparation of platinum nanotubes. Precursor for FePt superlattice magnetic nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15170-57-7
Molecular Formula C10H14O4Pt
Molecular Weight (g/mol) 393.30
MDL Number MFCD00000028
InChI Key KLFRPGNCEJNEKU-FDGPNNRMSA-L
Synonym platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii
PubChem CID 10960186
SMILES [Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 250°C to 252°C
Color Red
Physical Form Powder
Quantity 1 g
Solubility Information Very soluble in chloroform; insoluble in water
IUPAC Name platinum(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Formula Weight 393.31
Chemical Name or Material Platinum(II) 2, 4-pentanedionate, Pt 48.0% min.
Hazard Category H315-H319-H335-H361
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 239-223-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.