Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(II) 2,4-pentanedionate, Pt 48.0% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1052603

Quantity 1g

 View more versions of this product

Catalog No. AA1052603


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

15170-57-7
C10H14O4Pt
MFCD00000028
Platinum(II) 2, 4-pentanedionate, Pt 48.0% min.
Needles
393.31
Platinum(II) acetylacetonate

Specifications

15170-57-7
C10H14O4Pt
MFCD00000028
Red
Platinum(II) 2, 4-pentanedionate, Pt 48.0% min.
Needles
393.31
Platinum(II) acetylacetonate
1g

Safety and Handling

SDS & Safety Documents

H301-H332-H315-H319-H317: Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction.

EINECSNumber : 239-223-5

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit