Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(II) acetylacetonate, 98%

Catalog No. AC369350050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

Catalog No. AC369350050

Supplier: Thermo Scientific Chemicals 369350050

Only null left
Add to Cart

CAS: 15170-57-7 | C10H14O4Pt | 393.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15170-57-7
Molecular Formula C10H14O4Pt
Molecular Weight (g/mol) 393.30
MDL Number MFCD00000028
InChI Key KLFRPGNCEJNEKU-FDGPNNRMSA-L
Synonym platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii
PubChem CID 10960186
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;platinum
SMILES [Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 250.0°C to 252.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Pt)
Packaging Glass bottle
Linear Formula [CH3COCH=C(CH3)O]2Pt
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, diethyl ether;, soluble in acetone, toluene, diluted acids
Sublimation Point 170 °C
Formula Weight 393.31
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Platinum(II) acetylacetonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Cal
EINECSNumber 239-223-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.