Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(II) bromide, 98%

Catalog No. ac195410010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

CAS: 13455-12-4 | Br2Pt | 354.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-12-4
Molecular Formula Br2Pt
Molecular Weight (g/mol) 354.89
MDL Number MFCD00011180
InChI Key KGRJUMGAEQQVFK-UHFFFAOYSA-L
Synonym platinum ii bromide, platinum bromide, platinum bromide ptbr2, platinum dibromide, ptbr2, acmc-1c2hr, platinum bromide, ptbr2
PubChem CID 83486
IUPAC Name dibromoplatinum
SMILES [Br-].[Br-].[Pt++]

Specifications

Melting Point 250°C
Color Brown to Khaki
Physical Form Powder
Quantity 1 g
Assay Percent Range 97.5% min
Linear Formula PtBr2
Formula Weight 354.9
Percent Purity 98%
Packaging Glass bottle
Specific Gravity 6.65
Density 6.6500g/mL
Chemical Name or Material Platinum(II) bromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 236-648-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.