Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(II) bromide, 99.9% (metals basis), Pt 54.5% min

Catalog No. p-7048948
Click to view available options
Quantity:
1 g
5 g

CAS: 13455-12-4 | Br2Pt | 354.89 g/mol

Platinum(II) bromide used as Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-12-4
Molecular Formula Br2Pt
Molecular Weight (g/mol) 354.89
MDL Number MFCD00011180
InChI Key KGRJUMGAEQQVFK-UHFFFAOYSA-L
Synonym platinum ii bromide, platinum bromide, platinum bromide ptbr2, platinum dibromide, ptbr2, acmc-1c2hr, platinum bromide, ptbr2
PubChem CID 83486
IUPAC Name dibromoplatinum
SMILES [Br-].[Br-].[Pt++]

Specifications

Melting Point 250°C (decomposition)
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 354.91
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 54.5% min.
Assay (metals basis)
Chemical Name or Material Platinum(II) bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-648-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.