Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis)

Catalog No. p-7051702
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm

Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis) is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Iridium foil
Name Note 0.1mm (0.004 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
Quantity 25 x 25 mm
Odor Odorless
Weight ≈1.35g/25x25mm
Form Foil
Assay Percent Notes (metals basis)
MDL Number MFCD02091683
Solubility Information Insoluble in water.
Percent Purity 99.9%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.