Alfa Aesar™ Platinum Iridium gauze, 150 mesh woven from 0.043mm (0.0017 in.) dia. wire

Filtration, electrodes

Manufacturer: Alfa Aesar™ 40934FI

Quantity 50x50mm

 View more versions of this product

Catalog No. AA40934FI

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD02091683
    Chemical Name or Material Platinum Iridium gauze, Woven from 0.043mm (0.0017 in.) dia. wire

    Specifications

    Mesh Size 150 Mesh
    Odor Odorless
    Quantity 50x50mm
    TSCA TSCA
    Physical Form Wire Cloth
    MDL Number MFCD02091683
    Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
    Chemical Name or Material Platinum Iridium gauze, Woven from 0.043mm (0.0017 in.) dia. wire

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit