Learn More
Platinum Iridium wire, 0.127mm (0.005in) dia, annealed, 99.9% (metals basis excluding precious metals)
Description
Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes.Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes
Specifications
Specifications
| Chemical Name or Material | Platinum Iridium wire |
| Name Note | 0.127mm (0.005 in.) dia., Annealed, Temper: soft |
| Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 90:10 wt% |
| Quantity | 50 cm |
| Odor | Odorless |
| Weight | ≈0.03g/10cm |
| Form | Wire |
| Assay Percent Notes | (metals basis excluding precious metals) |
| MDL Number | MFCD02091683 |
| Solubility Information | Insoluble in water and acid. |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.