Learn More
Platinum Iridium wire, 0.5mm (0.02in) dia, hard
Description
Fabrication of metal microelectrodes for electrical stimulation of nervous tissue and electrophysiological recordings. Inoculating needles and leadwires for implants. Electrode material for gel boxes/electrophoresis chambers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Platinum Iridium wire |
| Name Note | 0.5mm (0.02 in.) dia., Hard, Temper: as drawn |
| Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 80:20 wt% |
| Quantity | 50 cm |
| Odor | Odorless |
| Weight | ≈0.44g/10cm |
| Form | Wire |
| MDL Number | MFCD02091683 |
| Solubility Information | Insoluble in water |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.