Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min

Catalog No. p-4407706
Click to view available options
Quantity:
1 g
5 g

CAS: 13454-96-1 | Cl4Pt | 336.88 g/mol

Platinum(IV) chloride is used as a catalyst and analytical reagent. Further, it is used in the production of semiconductor epitaxy and in electroplating process. It is involved in the preparation of platinum(II) chloride .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-96-1
Molecular Formula Cl4Pt
Molecular Weight (g/mol) 336.88
MDL Number MFCD00011182
InChI Key FBEIPJNQGITEBL-UHFFFAOYSA-J
Synonym platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4
PubChem CID 26031
IUPAC Name tetrachloroplatinum
SMILES Cl[Pt](Cl)(Cl)Cl

Specifications

Melting Point 370°C (decomposition)
Color Red to Brown
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water and acetone. Slightly soluble in ethanol and insoluble in ether.
Formula Weight 336.9
Sensitivity Hygroscopic
Density 4.303
Chemical Name or Material Platinum(IV) chloride, Pt 57% min.
Hazard Category H301-H314-H317-H334
Precautionary Statement P260-P264b-P270-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: II
EINECSNumber 236-645-1
RTECSNumber TP2275500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.