Alfa Aesar™ Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min

Manufacturer: Alfa Aesar™ 4450306

Quantity 5g

 View more versions of this product

Catalog No. AA4450306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13454-96-1
    Molecular Formula PtCl4
    Formula Weight 336.9
    MDL Number MFCD00011182
    UN Number UN2923
    Chemical Name or Material Platinum (IV) chloride
    Grade Premion™

    Specifications

    Formula Weight 336.9
    Health Hazard 2 GHS H Statement
    H301-H334-H314-H318-H317
    Toxic if swallowed.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 13454-96-1
    Health Hazard 1 Danger
    Density 4.303g/mL
    TSCA TSCA
    Odor Odorless
    Physical Form Crystalline Soluble
    Sensitivity Hygroscopic
    Color Brown
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Molecular Formula PtCl4
    MDL Number MFCD00011182
    Melting Point 370°C (dec.)
    Concentration Pt 57% min
    UN Number UN2923
    Quantity 5g
    RTECS Number TP2275500
    Chemical Name or Material Platinum (IV) chloride
    Assay ≥99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H301-H334-H314-H318-H317
    Toxic if swallowed.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit