Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min

Catalog No. p-7051075
Click to view available options
Quantity:
0.25 g
1 g
5 g

CAS: 1314-15-4 | O2Pt | 227.08 g/mol

Platinum(IV) oxide is used as catalyst for hydrogenation reactions. It is applied in the hydrogenation of alkenes to alkanes, nitro compounds to amines and ketones to alcohols. It is also involved in the hydrogenolysis of cyclopropyl group in to an isopropyl group and in selective oxidation of primary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-15-4
Molecular Formula O2Pt
Molecular Weight (g/mol) 227.08
MDL Number MFCD00011184
InChI Key YKIOKAURTKXMSB-UHFFFAOYSA-N
IUPAC Name dioxoplatinum
SMILES O=[Pt]=O

Specifications

Melting Point 450°C
Color Brown
Physical Form Powder
Quantity 0.25 g
Linear Formula PtO2
UN Number UN1479
Merck Index 14,7527
Solubility Information Soluble in caustic potash solution. Insoluble in water,acid,aquaregia.
Formula Weight 227.09
Percent Purity 99.95%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pt 84.4% min.
Odor Odorless
Density 10.2 g/mL
Assay (metals basis)
Chemical Name or Material Platinum(IV) oxide, Anhydrous
Show More Show Less
Hazard Category H271
Hazard Statement GHS H Statement
H272
May intensify fire
oxidizer.
Precautionary Statement P210-P220-P221-P280-P283-P306+P360-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: I
EINECSNumber 215-223-0
RTECSNumber TP2506020
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.