Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis)

Catalog No. AC211930010
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

CAS: 12137-21-2 | H2O3Pt | 245.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12137-21-2
Molecular Formula H2O3Pt
Molecular Weight (g/mol) 245.10
MDL Number MFCD00066964
InChI Key SPGAMGILENUIOF-UHFFFAOYSA-N
Synonym dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o
PubChem CID 11139377
SMILES O.O=[Pt]=O

Specifications

Color Brown to Black
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.8%
Linear Formula PtO2·H2O
Fieser 01,890
Merck Index 10, 7398
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in conc.acids and in dil.sol. of koh
IUPAC Name dioxoplatinum hydrate
Formula Weight 245.1
Percent Purity 99.80%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Platinum(IV) oxide monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.