Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
12137-21-2
O2Pt·H2O
245.1
MFCD00066964
SPGAMGILENUIOF-UHFFFAOYSA-N
dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o
11139377
dioxoplatinum;hydrate
O.O=[Pt]=O
12137-21-2
O2Pt·H2O
245.1
MFCD00066964
SPGAMGILENUIOF-UHFFFAOYSA-N
dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o
11139377
dioxoplatinum;hydrate
O.O=[Pt]=O
for ultra trace analysis
Assay Percent Range 99.8%
Linear Formula PtO2·H2O
Fieser 01,890
Merck Index 10, 7398
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in conc.acids and in dil.sol. of koh
Formula Weight 245.1
Physical Form Powder
Percent Purity 99.80%
Grade Trace Metal Basis
Packaging Glass bottle
Platinum (Pt) 77.6 to 82%
Solubility Solubility in water: insoluble in water. Other solubilities: soluble in conc.acids and in dil.sol. of koh
Surface Area 80 m2/g min.
Total Trace Metal Impurities 2000ppm max
Color Brown to Black
Quantity 250mg
Chemical Name or Material Platinum(IV) oxide monohydrate
Show More Show Less

GHS H Statement
May cause fire or explosion; strong oxidizer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger