Alfa Aesar™ Platinum(IV) oxide monohydrate, Adams Catalyst, Pt 75% min

Manufacturer: Alfa Aesar™ 1227806

Quantity 5g

 View more versions of this product

Catalog No. AA1227806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12137-21-2
    Linear Formula PtO2·H2O
    Formula Weight 245.10 (227.09 Anhydrous)
    MDL Number MFCD00066964
    UN Number UN1479
    Merck Index 14,7527
    Synonym Adams' Catalyst
    Chemical Name or Material Platinum(IV) oxide monohydrate, Adams Catalyst

    Specifications

    Formula Weight 245.10 (227.09 Anhydrous)
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P280-P370+P378a-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Dark brown to black
    Health Hazard 2 GHS H Statement
    H272
    May intensify fire
    oxidizer.
    RTECS Number TP2506020
    Physical Form Powder
    UN Number UN1479
    Synonym Adams' Catalyst
    Health Hazard 1 Danger
    Concentration Pt 75% min.
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    TSCA TSCA
    Linear Formula PtO2·H2O
    Surface Area >85m2/g
    MDL Number MFCD00066964
    CAS 12137-21-2
    Merck Index 14,7527
    Melting Point 450°C
    Quantity 5g
    Solubility Information Insoluble in water, acid, aqua regia
    Chemical Name or Material Platinum(IV) oxide monohydrate, Adams Catalyst

    Description

    • Hydrogenation catalyst
    • Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups

    GHS H Statement
    H272
    May intensify fire
    oxidizer.

    GHS P Statement
    P221-P210-P220-P280-P370+P378a-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit