Alfa Aesar™ Platinum(IV) sulfide, Pt 74.8% min

Manufacturer: Alfa Aesar™ 1399203

Quantity 1g

 View more versions of this product

Catalog No. AA1399203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12038-21-0
    Formula Weight 259.22
    MDL Number MFCD00016293
    Chemical Name or Material Platinum(IV) sulfide

    Specifications

    Formula Weight 259.22
    Physical Form Powder
    TSCA TSCA
    MDL Number MFCD00016293
    CAS 12038-21-0
    Concentration Pt 74.8% min.
    Melting Point 225° to 250°C (dec.)
    Quantity 1g
    Chemical Name or Material Platinum(IV) sulfide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit