Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum, 3% on activated carbon powder, standard, reduced, nominally 50% water wet

Catalog No. p-4824876
Click to view available options
Quantity:
5 g
25 g

Form: 900 - 1000m2 Standard, reduced, nominally 50% water wet | CAS: 7440-6-6

Platinum is used in catalytic converters, laboratory equipment, electrical application and electrodes, platinum resistance thermometers, dentistry equipment, and jewellery. It is also used in glass fiber applications, chemical processing, off-road vehicles and fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-6-6
MDL Number MFCD00011179

Specifications

Color Black
Quantity 5 g
Solubility Information Soluble in aqua regia. Insoluble in water and acid.
Odor Odorless
Appearance Black powder
Chemical Name or Material Platinum, 3% on activated carbon powder
Hazard Category H228
Precautionary Statement P210-P240-P241-P280-P370+P378q
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.