Alfa Aesar™ Platinum powder, -200 mesh, 99.98% (metals basis)

Manufacturer: Alfa Aesar™ 1207577

Quantity 0.1g

 View more versions of this product

Catalog No. AA1207577

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-06-4
    MDL Number MFCD00011179
    Chemical Name or Material Platinum powder

    Specifications

    Mesh Size −200 Mesh
    Physical Form Powder
    MDL Number MFCD00011179
    RTECS Number TP2160000
    TSCA TSCA
    Odor Odorless
    CAS 7440-06-4
    Quantity 0.1g
    Chemical Name or Material Platinum powder
    Assay 99.98%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit