Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum, 5% on alumina powder, reduced

Catalog No. AA4422214
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AA4422214

Supplier: Thermo Scientific Chemicals 04422214

Only null left
Add to Cart

CAS: 7440-6-6

Platinum on Alumina (aluminum oxide) is catalyst material with applications in synthetic chemistry such as hydrogenation of functional groups. catalyst is recommended for selective hydrogenation reactions. The particle size of the catalyst is ideal for allowing fast separation from the reaction mixture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-6-7440
MDL Number MFCD03458043

Specifications

Appearance Powder
Quantity 25 g
Form Powder
Solubility Information Insoluble in water,moderately soluble in strong mineral acids.
Chemical Name or Material Platinum, 5% on alumina powder
Hazard Category H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.