Alfa Aesar™ Platinum, 5% on alumina powder, reduced, Alfa Aesar™

Manufacturer: Alfa Aesar™ 4422222

Quantity 100g

 View more versions of this product

Catalog No. AA4422222

  Check Availability
Add to Cart

    Specifications

    Quantity 100g
    Physical Form Powder
    MDL Number MFCD00011179
    Health Hazard 1 H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Appearance Powder
    Chemical Name or Material Platinum, 5% on alumina powder

    H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit