Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ Taq DNA Polymerase, High Fidelity

Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required.

Supplier:  Invitrogen™ 11304102

Encompass_Preferred

 View more versions of this product

Catalog No. 11-304-102


Only null left
Add to Cart
Request a Quote

Includes: Platinum Taq DNA Polymerase High Fidelity (1000μL, at 5U/μL) is supplied with a bottle (50mL) of 10X High Fidelity Buffer (600mM Tris-SO4 (pH 8.9), 180mM (NH4)2SO4), and a bottle (25mL) of 50mM MgSO4.

Description

Description

Platinum Taq DNA Polymerase, High Fidelity, is ideal for amplification of DNA fragments when high yields and robust amplification are required. High fidelity is provided by a mixture of Platinum Taq DNA Polymerase and the proofreading enzyme. PCR specificity is improved with built-in Platinum hot-start technology.

Features of Platinum Taq DNA Polymerase, High Fidelity

  • Greater than six times higher fidelity than Taq DNA polymerase
  • Amplification of fragments up to 15 kb
  • Convenient room temperature reaction assembly
  • Increased specificity, and yield

Applications

  • Amplification of complex DNA templates
  • Amplification of viral and plasmid templates
  • RT-PCR

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Platinum Taq DNA Polymerase High Fidelity (1 x 1000 μL at 5 U/μL)
• 10X High Fidelity Buffer (1 x 50 mL)
• 50 mM MgSO4 (1 x 25 mL)

Store at -10°C to -30°C.
Low
Platinum Taq DNA Polymerase High Fidelity
DNA Polymerase
Dry Ice
6X
5000 Reactions
Separate Components
Primer-probe
1-step RT-qPCR
Standard
5,000 reactions
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Mixed, 3'-A
20 kb or less
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.