Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Platinum Iridium wire, 0.5mm (0.02in) dia, hard

Catalog No. p-4824882
Click to view available options
Quantity:
10 cm
50 cm
250 cm

Form: Approx. 0.44g/10cm, Temper: as drawn | CAS: 7440-6-6 | Pt:Ir; 90:10 wt%

Fabrication of metal microelectrodes for electrical stimulation of nervous tissue and electrophysiological recordings. Inoculating needles and leadwires for implants. Electrode material for gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-6-6
Molecular Formula Pt:Ir; 90:10 wt%
MDL Number MFCD02091683

Specifications

Color White
Appearance Silvery white wire
Quantity 10 cm
Odor Odorless
Form Wire
Solubility Information Insoluble in water
Chemical Name or Material Platinum, Hard
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.