Platinum Wire, 0.5mm (0.02 in.) dia., hard, , Alfa Aesar™

Form: Approx. 0.44g/10cm, Temper: as drawn

Manufacturer: Alfa Aesar 10056BS

Quantity 10cm

 View more versions of this product

Catalog No. AA10056BS

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Wire, 0.5mm (0.02 in.) dia.
    Molecular Formula Pt:Ir; 90:10 wt%
    MDL Number MFCD02091683
    Chemical Name or Material Platinum, Hard

    Specifications

    Quantity 10cm
    Color Silvery white
    Physical Form Wire, 0.5mm (0.02 in.) dia.
    Molecular Formula Pt:Ir; 90:10 wt%
    MDL Number MFCD02091683
    Odor Odorless
    Appearance Silvery white wire
    Chemical Name or Material Platinum, Hard

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit