Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLAUR Mouse anti-Human, Unconjugated, Clone: 2D6, Abnova™

Catalog No. 89339180 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-180 Supplier Abnova Corporation Supplier No. H00005329M02
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

PLAUR Monoclonal antibody specifically detects PLAUR in Human samples. It is validated for ELISA, WB-Re.

Specifications

Antigen PLAUR
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2D6
Conjugate Unconjugated
Description plasminogen activator, urokinase receptor
Gene PLAUR
Gene Accession No. NM_002659
Gene Alias CD87, UPAR, URKR
Host Species Mouse
Immunogen PLAUR (NP_002650, 25 a.a. ∼ 134 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 5329
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.