Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Playing with Polymers
GREENER_CHOICESCI_ED

Catalog No. S02008 Shop All Aldon Products
Change view
Click to view available options
Grade:
K to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02008 K to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING CHOKING HAZARD Small parts. Not for children under 3 yrs.
Catalog No. S02008 Supplier Innovating Science™ Supplier No. IS1002
Add to Cart
Add to Cart
WARNING CHOKING HAZARD Small parts. Not for children under 3 yrs.

Students will learn about polymers are used in our everyday lives and make two different polymers - worms and slime.

Teacher leads a class discussion about polymers and their importance in our everyday lives (specifically plastics). After the discussion students will be able to make two different polymers - worms and slime. The activity contains enough materials for 6 groups of students

Specifications

Grade K to 5
Class Size 6 Lab Groups
Product Type Properties of Polymers Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.