Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120091
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120091 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120091 Supplier Invitrogen™ Supplier No. PA5120091
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-431, U-87MG, HL-60, HT-29, Mouse testis, Mouse lung, Mouse skeletal muscle, Rat testis. The target is usually found in the following locations: Apical cell membrane, Single-pass type I membrane protein. Immunogen sequence: ARPNNSSDLP TSWRGLSWSI GGDGNLETHT TLPNILKKFN PYLLGFSTST WEGTAGLNVA AEGARARDMP AQAWDLVERM KNSPDINLEK DWKLVTLFIG VNDLCHYCEN PEAHLATEYV QHIQQALDIL SEELPRAFVN VVEVMELASL YQGQGGKCAM LAAQNNCTCL RHSQSSLEKQ ELKKVNWNLQ HGISSFSYWH QYTQREDFAV VVQPFFQNTL TPLNERGDTD LTFFSEDCFH FSDRGHAEMA IALWNNMLEP VGRKTTSNNF THSRAKLKCP SPESPYLYTL RNSRLLPDQA EEAPEV.

This gene encodes a membrane-associated phospholipase that displays lysophospholipase and phospholipase A2 activities through removal of sn-1 and sn-2 fatty acids of glycerophospholipids. In addition, it displays lipase and retinyl ester hydrolase activities. The encoded protein is highly conserved and is composed of a large, glycosylated extracellular domain composed of four tandem homologous domains, followed by a hydrophobic segment that anchors the enzyme to the membrane and a short C-terminal cytoplasmic tail. This gene has been identified as a candidate rheumatoid arthritis risk gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLB1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLB1
Gene Accession No. O54728, Q3TTY0, Q6P1J6
Gene Alias 4632413E21Rik; 4930433E17Rik; 4930539A06Rik; BC033606; hPLB; Lysophospholipase; Phlpb; Phospholipase A2; phospholipase B; Phospholipase B/lipase; phospholipase B1; phospholipase B1, membrane-associated; PLB; PLB/LIP; PLB1
Gene Symbols PLB1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151056, 192259, 665270
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.