Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLBD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568857
Description
This target displays homology in the following species: Cow: 77%; Dog: 86%; Guinea Pig: 82%; Horse: 79%; Human: 79%; Mouse: 100%; Pig: 86%; Rabbit: 79%; Rat: 92%; Zebrafish: 82%.
PLBD1 may act as an amidase or a peptidase that works with the small size of a putative binding pocket. PLBD1 exhibits a weak phospholipase activity, acting on various phospholipids, including phosphatidylcholine, phosphatidylinositol, phosphatidylethanolamine and lysophospholipids.
Specifications
PLBD1 | |
Polyclonal | |
Unconjugated | |
PLBD1 | |
1100001H23Rik; hypothetical protein; im:7138859; LAMA-like protein 1; Lamina ancestor homolog 1; phospholipase B domain containing 1; phospholipase B domain-containing protein 1; Phospholipase B-like 1; Phospholipase B-like 1 chain A; Phospholipase B-like 1 chain B; Phospholipase B-like 1 chain C; PLB homolog 1; PLBD1; putative phospholipase B-like 1; RGD1308734; zgc:194816 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
66857 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8VCI0 | |
PLBD1 | |
synthetic peptide corresponding to a region of mouse PLBD1 (aa 51-100). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction