Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLBD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559137
Description
Immunogen sequence: WMPAEKTVQV KNVMDKNGDA YGFYNNSVKT TGWGILEIRA GYGSQTLSNE IIMFVAGFLE GYLTAPHMND HYTNLYPQLI TKPSIMDKVQ DFMEKQD Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.
PLBD1 may act as an amidase or a peptidase that works with the small size of a putative binding pocket. PLBD1 exhibits a weak phospholipase activity, acting on various phospholipids, including phosphatidylcholine, phosphatidylinositol, phosphatidylethanolamine and lysophospholipids.
Specifications
| PLBD1 | |
| Polyclonal | |
| Unconjugated | |
| PLBD1 | |
| 1100001H23Rik; hypothetical protein; im:7138859; LAMA-like protein 1; Lamina ancestor homolog 1; phospholipase B domain containing 1; phospholipase B domain-containing protein 1; Phospholipase B-like 1; Phospholipase B-like 1 chain A; Phospholipase B-like 1 chain B; Phospholipase B-like 1 chain C; PLB homolog 1; PLBD1; putative phospholipase B-like 1; RGD1308734; zgc:194816 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79887 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P4A8 | |
| PLBD1 | |
| Recombinant protein corresponding to Human PLBD1. Recombinant protein control fragment (Product #RP-102885). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction