Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLBD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553557 Supplier Invitrogen™ Supplier No. PA553557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDWFSYDGSP RAQIFRRNQS LVQDMDSMVR LMRYNDFLHD PLSLCKACNP QPNGENAISA RSDLNPANGS YPFQALRQRS HGGIDVKVTS MSLARILSLL AASGPTWDQV PPFQWSTS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.

PLBD2 (Phospholipase B Domain Containing 2) is a Protein Coding gene. Diseases associated with PLBD2 include Gallbladder Papillomatosis and Gm1-Gangliosidosis, Type Iii. Gene Ontology (GO) annotations related to this gene include hydrolase activity. An important paralog of this gene is PLBD1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLBD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLBD2
Gene Accession No. Q8NHP8
Gene Alias 1300012G16Rik; 66.3 kDa protein; 76 kDa protein; AU019810; LAMA-like protein 2; Lamina ancestor homolog 2; mannose-6-phosphate protein associated protein p76; mannose-6-phosphate protein p76; p76; phospholipase B domain containing 2; phospholipase B domain-containing protein 2; phospholipase B-like 2 32 kDa form; phospholipase B-like 2 45 kDa form; PLB homolog 2; Plbd2; Putative phospholipase B-like 2; Putative phospholipase B-like 2 15 kDa form; Putative phospholipase B-like 2 28 kDa form; Putative phospholipase B-like 2 32 kDa form; Putative phospholipase B-like 2 40 kDa form; Putative phospholipase B-like 2 45 kDa form; RDCR-0918-3; RDCR-0918-3 protein
Gene Symbols PLBD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLBD2. Recombinant protein control fragment (Product #RP-90359).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196463
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.