Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLC-beta 3 Antibody (SR1198), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322249100UL
Description
PLC-beta 3 Monoclonal antibody specifically detects PLC-beta 3 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
PLC-beta 3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1198 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3, EC 3.1.4.11, FLJ37084, Phosphoinositide phospholipase C-beta-3, phospholipase C, beta 3 (phosphatidylinositol-specific), Phospholipase C-beta-3, PLC beta 3, PLC-beta-3 | |
A synthesized peptide derived from human PLC-beta 3 (Uniprot #: Q01970) | |
100 μg | |
Lipid and Metabolism, Phospho Specific | |
5331 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction