Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLC-gamma 2 Antibody (SR1168), Novus Biologicals™
SDP

Catalog No. NB260096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260096 50 μg
NB258818 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260096 Supplier Novus Biologicals Supplier No. NBP32224550UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PLC-gamma 2 Monoclonal antibody specifically detects PLC-gamma 2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen PLC-gamma 2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone SR1168
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias EC 3.1.4.11, Phosphoinositide phospholipase C-gamma-2, phospholipase C gamma 2, phospholipase C, gamma 2 (phosphatidylinositol-specific), phospholipase C, gamma 2 (phosphatidylyinositol-specific), Phospholipase C-gamma-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2, Phospholipase C-IV, PLCG2, PLC-gamma-2, PLC-IV
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLC-gamma 2 (Uniprot #: P16885)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5336
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.