Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLC gamma2 Polyclonal Antibody, Abbomax

Catalog No. 502203606
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3606 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3606 Supplier Abbomax Inc Supplier No. 50012474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Jurkat whole cell lysate Cellular location: Cell projection, lamellipodium, Cell projection, ruffle, Rapidly redistributed to ruffles and lamellipodia structures in response to epidermal growth factor (EGF) treatment.

PLC gamma2 Polyclonal antibody specifically detects PLC gamma2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PLC gamma2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Plcg2
Gene Accession No. P16885, P24135, Q8CIH5
Gene Alias 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2; APLAID; FCAS3; phosphoinositide phospholipase C; phosphoinositide phospholipase C-gamma-2; phospholipase C gamma 2; phospholipase C, gamma 2; phospholipase C, gamma 2 (phosphatidylinositol-specific); phospholipase C-gamma-2; Phospholipase C-IV; PLC gamma 2; PLC gamma2; PLC- gamma2; PLC gamma-2; PLCG2; Plcg-2; PLC-gamma; PLCgamma2; PLC-gamma-2; PLC-IV
Host Species Rabbit
Immunogen A synthetic peptide of human PLC gamma 2 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234779, 29337, 5336
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.