Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115767
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115767 Supplier Invitrogen™ Supplier No. PA5115767
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PLCB1.

The protein encoded by this gene catalyzes the formation of inositol 1,4,5-trisphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. This reaction uses calcium as a cofactor and plays an important role in the intracellular transduction of many extracellular signals. This gene is activated by two G-protein alpha subunits, alpha-q and alpha-11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLCB1
Gene Accession No. P10687, Q9NQ66, Q9Z1B3
Gene Alias 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-1; 3110043I21Rik; AI132408; EIEE12; inositoltrisphosphohydrolase; KIAA0581; mKIAA0581; monophosphatidylinositol phosphodiesterase; Phosphb; phosphoinositidase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-beta-1; phosphoinositide-specific phospholipase c; phospholipase C; phospholipase C beta 1; phospholipase C, beta 1; phospholipase C, beta 1 (phosphoinositide-specific); Phospholipase C-beta1; phospholipase C-beta-1; phospholipase C-beta-1a; phospholipase C-I; PI-PLC; PLC'1; PLC154; PLC-154; Plcb; PLCB1; PLCB1A; PLCB1B; PLCbeta1; PLC-beta-1; PLC-I; RATPHOSPHB; triphosphoinositide phosphodiesterase
Gene Symbols PLCB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLCB1(Accession Q9NQ66), corresponding to amino acid residues G449-P499.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18795, 23236, 24654
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.