Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLCB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526260
Description
PLCB2 Polyclonal Antibody for Western Blot, IHC (P), Flow
PLCB2 is the production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by activated phosphatidylinositol-specific phospholipase C enzymes.
Specifications
| PLCB2 | |
| Polyclonal | |
| Unconjugated | |
| PLCB2 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-2; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-2; AI550384; B230205M18Rik; B230399N12; Phosphoinositide phospholipase C-beta-2; phospholipase C beta 2; phospholipase C, beta 2; phospholipase C-beta-2; PKC beta II; PLC beta-2; PLCB2; plcb2 {ECO:0000250; PLCbeta2; PLC-beta2; PLC-beta-2; PLCbeta-2; UniProtKB:Q00722} | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 5330 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q00722 | |
| PLCB2 | |
| KLH conjugated synthetic peptide between 174-202 amino acids from the N-terminal region of human PLCB2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction