Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102006 Supplier Invitrogen™ Supplier No. PA5102006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PLCB2.

PLCB2 is the production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by activated phosphatidylinositol-specific phospholipase C enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLCB2
Gene Accession No. A3KGF7, O89040, Q00722
Gene Alias 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-2; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-2; AI550384; B230205M18Rik; B230399N12; Phosphoinositide phospholipase C-beta-2; phospholipase C beta 2; phospholipase C, beta 2; phospholipase C-beta-2; PKC beta II; PLC beta-2; PLCB2; plcb2 {ECO:0000250; PLCbeta2; PLC-beta2; PLC-beta-2; PLCbeta-2; UniProtKB:Q00722}
Gene Symbols PLCB2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLCB2(Accession Q00722), corresponding to amino acid residues S341-I391.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18796, 5330, 85240
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.