Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103068 Supplier Invitrogen™ Supplier No. PA5103068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PLCB3.

PLCB3 encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCB3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Plcb3
Gene Accession No. P51432, Q01970, Q99JE6
Gene Alias 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-3; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3; EC 3.1.4.11; mKIAA4098; Phosphoinositide phospholipase C-beta-3; phosphoinositide-specific phospholipase c; phospholipase C beta 3; phospholipase C, beta 3; phospholipase C, beta 3 (phosphatidylinositol-specific); phospholipase C-beta-3; PLC beta 3; Plcb3; PLCbeta3; PLC-beta3; PLC-beta-3
Gene Symbols Plcb3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLCB3(Accession Q01970), corresponding to amino acid residues T1078-H1128.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18797, 29322, 5331
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.