Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567835 Supplier Invitrogen™ Supplier No. PA567835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

The production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by activated phosphatidylinositol-specific phospholipase C enzymes. This form has a role in retina signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCB4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLCB4
Gene Accession No. Q15147, Q9QW07
Gene Alias 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-4; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-4; A930039J07Rik; AI854601; ARCND2; Beta4aa; C230058B11Rik; dJ1119D9.2 (Phopholipase C, beta 4 (1-Phosphatidylinositol-4,5-Bisphosphate Phosphodiesterase Beta 4)); inositoltrisphosphohydrolase; monophosphatidylinositol phosphodiesterase; PCL-C1; phosphoinositidase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-beta-4; phosphoinositide-specific phospholipase c; phospholipase C; phospholipase C beta 4; phospholipase C, beta 4; phospholipase C-beta-4; PI-PLC; PLCB4; PLC-beta-4; RATBETA4AA; triphosphoinositide phosphodiesterase
Gene Symbols PLCB4
Host Species Rabbit
Immunogen Synthesized peptide derived from human PLCB4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18798, 25031, 5332
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.