Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLCD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554151
Description
Immunogen sequence: EAFYKMLTQR VEIDRTFAEA AGSGETLSVD QLVTFLQHQQ REEAAGPALA LSLIERYEPS ETAKAQRQMT KDGFLMYLLS ADGSAFSLAH RRVYQDMGQP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
Phosphoinositide-specific phospholipase C (PLC) acts as a signal transducer that generates 2 second messengers, diacylglycerol and inositol 1, 4, 5-trisphosphate, by hydrolyzing inositol phospholipids. PLC comprises a diverse family of enzymes that differ in structure and tissue distribution (Berridge, 1993 ).
Specifications
| PLCD1 | |
| Polyclonal | |
| Unconjugated | |
| PLCD1 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1; AW212592; C79986; NDNC3; phosphoinositide phospholipase C; phosphoinositide phospholipase C-delta-1; phospholipase C delta 1; phospholipase C delta 1 Lf; phospholipase C delta 1 long form; phospholipase C, delta 1; Phospholipase C-delta1; phospholipase C-delta-1; phospholipase C-III; Plc1; Plcd; PLCD1; PLC-delta 1; PLC-delta-1; PLC-III | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5333 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51178 | |
| PLCD1 | |
| Recombinant protein corresponding to Human PLCD1. Recombinant protein control fragment (Product #RP-92493). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction